MaxiFlex® Endurance™ Seamless Knit Nylon Gloves, Protective Industrial Products
Supplier: PIP
34-844/S 34-844/M 34-844/L 34-844/XL 34-845/S 34-845/M 34-845/L 34-845/XL 34-844/S 34-844/M 34-844/L 34-844/XL 34-845/S 34-845/M 34-845/L 34-845/XL
20002-368CS 3511.08 CAD
20002-368 20002-370 20002-372 20002-374 20002-376 20002-378 20002-380 20002-382 75814-428 75814-430 75814-432 75814-434 75814-438 75814-440 75814-442 75814-444
MaxiFlex® Endurance™ Seamless Knit Nylon Gloves, Protective Industrial Products
Gloves
Designed and developed as a breathable glove, MaxiFlex® Endurance™ incorporates raised micro dots to increase durability and offer more cushioning in highly repetitive applications requiring precision handling.
  • Gloves incorporate unique Micro Dots on palm for excellent grip
  • Ultra cool and comfortable to wear
  • Excellent dexterity, sensitivity, and tactility
  • Designed to produce minimal lint and dust
  • Minimal penetration keeps inner liner soft
With applications in general assembly, micro-engineering, automotive components, painting, and horticulture, these gloves offer a wide array of uses in diverse applications. The micro foam nitrile coatings are compatible with light oils and will provide a good grip and excellent abrasion resistance. Knit wrists help prevent dirt and debris from entering the glove. The patented micro foam nitrile coating offers 360° breathability, and the 'hand at rest' design reduces hand fatigue and increases comfort. Gloves are washable in tempered water not exceeding 40 °C (104 °F). Gloves are available in two nitrile coating coverage options.
Certifications: CFIA approved. Component material complies with Federal regulations for food contact, 21 CFR, 170-199.
Packaging: Gloves are packaged 12 per pack, 12 dozen per case.