Kleinert-Kutz Rongeur, OR Grade, Sklar®
Supplier: Sklar
40-4190 40-4192 40-4194
10195-908EA 1503.6 CAD
10195-908 10195-910 10195-912
Kleinert-Kutz Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Kleinert-Kutz rongeur is a double action, orthopedic instrument used in the synovial joint in treatment of rheumatoid arthritis.
  • Available in different jaw curvatures and bite sizes
  • Made of stainless steel
  • Industry standard
  • Premium OR-Grade
The curvature mirrors the roundedness of the joint for a firm grasp without ripping the synovium.