Gigli Saw Wire, OR Grade, Sklar®
Supplier: Sklar
40-5542
10163-404EA 43.62 CAD
10163-404
Gigli Saw Wire, OR Grade, Sklar®
Wire
Sklar's® Gigli Saw Wire is comprised of multiple, flexible, wires twisted in such a way as to create multiple cutting surfaces.
The wire is attached to two Gigli Saw Handles, either of the T-shape or Loop shape, and moved back and forth about 180 degrees around the bone.
It is a single use piece of the saw due to the amounts of flesh and bone imbedded in the wire during procedures.