Gelpi Retractor, OR Grade, Sklar®
Supplier: Sklar
22-8850 22-8853 22-8855 60-6570 60-6580 22-8851
82030-154EA 697 CAD
82030-154 82030-156 82030-158 10197-826 10197-828 76192-932
Gelpi Retractor, OR Grade, Sklar®
Retractors
The Sklar® Gelpi Retractor is a ratcheted, finger ring retractor often used in smaller surgical sites.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
A single sharp prong on each tip is turned outward, sideways, at a 90° angle. Once the targeted tissue has been held back, the locking mechanism allows the retractor to remain in place, and does not require someone to hold it during the procedure.