Cottle Septal Speculum, OR Grade, Sklar
Supplier: Sklar
41-1194 41-1192 41-1190 41-1196
10194-058EA 644.19 CAD
10194-058 10194-304 10199-386 10199-396
Cottle Septal Speculum, OR Grade, Sklar
Specula
Sklar's Cottle Septal Speculum is used to provide internal visualization of the nasal valve, the most narrow area of the airway.
  • Speculum has a screw which enables it to be self-retaining
It features thin, narrow blades used for retracting nasal walls to allow visualization of nasal passage. This is necessary for rhinoplasty and other plastic surgeries as well as procedures such as a correcting a deviated septum.