Zalkind Ribbon Retractor, OR Grade, Sklar
Supplier: Sklar
22-8067 22-8075 60-3594 60-3596 60-3598 60-3607 60-3610 60-3612 60-3615 60-3617 60-3620 60-3625 60-3630
89025-396EA 220.25 CAD
89025-396 89025-398 10197-734 10197-736 10197-738 10197-740 10197-742 10197-744 10197-746 10197-748 10197-750 10197-752 10197-754
Zalkind Ribbon Retractor, OR Grade, Sklar
Retractors
Zalkind Ribbon Retractor is a handheld, malleable retractor useful in multiple procedures and surgical sites. It is available in different lengths and widths to accommodate regions of the body.
The malleability allows this retractor to be adjusted to adequately and safely retract a variety of tissue and organs to keep them from obstructing the surgeon's view and cluttering the surgical site
Censitrac Ready products are industry standard and constructed of stainless steel.
Smaller ribbon retractors may be used in neurosurgical or ophthalmic procedures, Larger retractors may be used in abdominal or cardiovascular procedures to hold back larger organs. They may also be used during body contouring to insert implants.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.