Drosophila Diet, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
ID001-1L ID001-5L
CA95060-286EA 715.35 CAD
CA95060-286 CA95060-288
Drosophila Diet, HiMedia
Cell Culture Supplements and Additives
Drosophila diet, ID001 is a modification of E
  • Modification of E. B. Lewis's original formula
  • Mimics the insect's normal food source
  • Provides the necessary nutrition for growth and development of fruit fly larvae
This medium mimics the insect's normal food source and provides the necessary nutrition for growth and development of fruit fly larvae.
B. Lewis's original formula for fruit fly larvae.