Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
Supplier: Corning
99999-15 99999-18 99999-13
60826-483CS 407.66 CAD
60826-483 60826-494 60827-110
Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
Closures Test/Sample Tube Closures
These disposable Corning® caps are made from phenolic resin and are autoclavable at 121°C
  • Available in different sizes
They are provided with a white rubber liner.