PYREX® Bomb for Gum Stability, Corning
Supplier: Corning
PYREX®
7732
52933-193CS 128.91 CAD
52933-193
PYREX® Bomb for Gum Stability, Corning
Petroleum Testing Apparatus
Bomb with cover made from PYREX® borosilicate glass to determine gum stability in gasoline.
  • Wide mouth with pourout style top
  • Cover to prevent condensed vapours from contaminating the sample
  • No interference with the free flow of oxygen
Replacement covers are available seperately.
Certifications: ASTM D-525