Corning® Polypropylene Funnel, Corning
Supplier: Corning
6120P-30 6120P-40 6120P-50 6120P-75 6120P-100 6120P-120 6120P-150
30617-106CS 145.79 CAD
30617-106 30617-108 30617-110 30617-112 30617-114 30617-116 30617-118
Corning® Polypropylene Funnel, Corning
Funnels Filtration Funnels Filter Funnels
Funnels, made of durable PP, translucent.
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Allow safe and easy transfers of solids or liquids from one vessel to another.