Iscove's Modified Dulbecco's Medium (IMDM) Mammalian Cell Culture Medium, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
AT070-5L AT070-20L
CA95016-506EA 89.54 CAD
CA95016-506 CA95016-508 CA95016-510
Iscove's Modified Dulbecco's Medium (IMDM) Mammalian Cell Culture Medium, HiMedia Laboratories
Cell Culture Media
Iscove’s Modified Dulbecco’s Medium is an enriched modification of Dulbecco’s Modified Eagle’s Medium wherein serum can be partially or totally replaced by chemically defined substances.
  • Contains additional amino acids, sodium selenite, sodium pyruvate, vitamins and inorganic salts
  • The first medium utilizing HEPES buffer
  • Supports good growth of precursor cells of erythrocytes and macrophages
  • Supports good growth of T and B lymphocytes
The medium contains additional amino acids, sodium selenite, sodium pyruvate, vitamins and inorganic salts. Potassium nitrate is substituted by ferric nitrate. IMDM was the first medium utilizing HEPES buffer. The medium when appropriately supplemented supports good growth of precursor cells of erythrocytes and macrophages. The medium also supports good growth of T and B lymphocytes and a variety of hybrid cells under serum free or reduced serum conditions.