PYREX® Rotary Evaporator Trap, with [ST] Inner and Outer Joints, Corning
Supplier: Corning
4000-25024 4000-10024
26307-270CS 434.41 CAD
26307-270 26307-262 26307-264
PYREX® Rotary Evaporator Trap, with [ST] Inner and Outer Joints, Corning
Vacuum Traps
These PYREX® traps are used in rotary evaporators. They have a round body with upper and lower stems.
  • Lower stem has a Standard Taper inner joint at the bottom and extends upward inside the body
  • The closed top of this stem has two side openings
  • The upper stem has a Standard Taper outer joint