PYREX® Liebig Condensers, with Drip Tip and [ST] Outer and Inner Joints, Corning
Supplier: Corning
2400-400 2400-200 2400-300
89091-374CS 1752.14 CAD
89091-374 89091-436 23132-088 89090-726
PYREX® Liebig Condensers, with Drip Tip and [ST] Outer and Inner Joints, Corning
Condensers
Made of PYREX® borosilicate glass.
  • With outer joint at the top and inner standard taper joint with drip tip
  • Very good temperature resistance
  • Very good chemical resistance
This general purpose condenser can be used for distillation, refluxing and extraction operations. The condenser tubulations are approximately 10 mm Ext.Ø. Can be used for vacuum distillation.