PYREX® Liebig Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
Supplier: Corning
2360-400 2360-500
89091-372CS 1588.44 CAD
89091-372 23132-044
PYREX® Liebig Condenser, with Drip Tip and 24/40 [ST] Inner Joint, Corning
Condensers
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This PYREX® general purpose condenser has a 24/40 Standard Taper inner joint at the bottom with a drip tip. Although the condensing area per unit length of the 400 mm long jacket is low, the large capacity water jacket gives efficient cooling. They can be used for either distillation or extraction. Condenser tubulations are approximately 10 mm Ext.Ø.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey