PYREX® Gas Washing Bottle with Plain Tip Tube, Corning
Supplier: Corning
1760-250 1760-500 1760-125
16429-043CS 558.26 CAD
16429-043 16429-065 16429-021
PYREX® Gas Washing Bottle with Plain Tip Tube, Corning
Bottles Gas Washing Bottles
Made of PYREX®, borosilicate glass.
  • Humidify carbon dioxide-air mixtures for maintaining high humidity
  • Large hexagonal base for stability
  • Side arm tubulations are 8 mm Ext.Ø
Gas washing bottles are used for washing and drying gases. NS 29/42 stopper is used to seal the top to the base and it incorporates a plain tip tube as the distributor.