PYREX® Gas Washing Bottles with Coarse Fritted Disc, Corning
Supplier: Corning
31760-500C 31760-125EC 31760-125C 31760-250C 31760-500EC 31760-500BO
16405-067CS 855.43 CAD
16405-067 16405-089 16405-023 16405-045 16405-125 16405-131
PYREX® Gas Washing Bottles with Coarse Fritted Disc, Corning
Bottles Gas Washing Bottles
PYREX®, borosilicate glass.
  • Large hexagonal base
  • Humidify carbon dioxide-air mixtures for maintaining high humidity
  • Fritted disc provides greater efficiency and more uniform dispersion of gas bubbles for complete absorption
These gas washing bottles are used for washing and drying gases. NS 40/50 stopper is used to seal the top to the base. The NS stopper incorporates a tube with a 20 mm diameter extra coarse fritted disc as the distributor. Tubulation outer dimension is 8 mm. This extra coarse frit has a nominal pore distribution of 170 to 220 µm.