Nutrient Mixture F-12 Ham Mammalian Cell Culture Medium, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
AT144-5L AT144-20L AT025-5L AT025-20L AT025-50L
CA89138-006EA 357.74 CAD
CA89138-006 CA89138-008 CA89138-010 CA89138-012 CA95016-618 CA95016-620 CA95016-622 CA95057-920
Nutrient Mixture F-12 Ham Mammalian Cell Culture Medium, HiMedia
Cell Culture Media
Ham’s Nutrient mixtures were originally developed for single cell plating of near diploid Chinese hamster ovary (CHO) cells and mouse L-cells.
  • Formulated for use with or without serum
  • Designed for serial propagation and cloning of two CHO cell lines
  • Medium of choice for the growth of cells of rodent origin
  • Also for cloning of myeloma and hybridoma cells
Both F-10 and F-12 are formulated for use with or without serum, depending on the type of cells being cultured. Ham’s nutrient mixture F12 was originally designed for serial propagation and cloning of two CHO cell lines namely, CHD-3 and CHL-1 and mouse L cells. It is the medium of choice for the growth of cells of rodent origin and for cloning of myeloma and hybridoma cells. This medium is also the medium of choice for clonal toxicity assay using CHO cells.