VWR® Dulbecco's Phosphate Buffered Saline (DPBS)
Supplier: VWR
02-0119-0500 02-0119-1000
VWRL0119-0500CS 373.76 CAD
VWRL0119-0500 VWRL0119-1000
VWR® Dulbecco's Phosphate Buffered Saline (DPBS)
Buffers Cell Culture Buffers
DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
  • Used to irrigate, wash, and dilute mammalian cells
  • Phosphate buffering maintains the pH in the physiological range
  • Available with or without calcium and magnesium
DPBS is used to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.