Nutrient Mixture F-10 Ham Mammalian Cell Culture Medium, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
AT024-5L AT024-20L
CA95016-612EA 249.13 CAD
CA95016-612 CA95016-614 CA95016-616
Nutrient Mixture F-10 Ham Mammalian Cell Culture Medium, HiMedia
Cell Culture Supplements and Additives
Ham’s Nutrient mixtures were originally developed for single cell plating of near diploid Chinese hamster ovary (CHO) cells and mouse L-cells.
  • Formulated for use with or without serum
  • Designed for clonal growth of CHO cells and chick embryo cells
  • Widely used for culturing a variety of cells
Both F-10 and F-12 are formulated for use with or without serum, depending on the type of cells being cultured. Ham’s nutrient mixture F10 was designed for clonal growth of CHO cells and chick embryo cells under serum free conditions. It is now widely used for culturing a variety of cells which include human diploid cells and white blood cells for chromosomal analysis and primary explants of rat, rabbit and chicken tissues.