Dulbecco's Modified Eagle Medium (DMEM) Mammalian Cell Culture Medium, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
AT006-1L AT006-5L AT006-20L AT007-1L AT007-5L AT007-20L AT066-1L AT066-5L AT066-20L AT067-1L AT067-5L AT067-20L AT068-1L AT068-5L AT068-20L AT065A-1L AT065A-5L AT065A-20L AT063A-20L
CA95016-456EA 126.64 CAD
CA95016-456 CA95016-458 CA95016-460 CA95016-462 CA95016-464 CA95016-466 CA95016-468 CA95016-470 CA95016-472 CA95016-474 CA95016-476 CA95016-478 CA95016-480 CA95016-482 CA95016-484 CA95016-486 CA95016-654 CA95016-656 CA95016-658 CA95057-914 CA95057-916
Dulbecco's Modified Eagle Medium (DMEM) Mammalian Cell Culture Medium, HiMedia
Cell Culture Media
DMEM is a modification of Basal Medium Eagle (BME) that contains four fold concentration of amino acids and vitamins.
  • One of the most widely used modification of Eagle’s medium
  • Includes glycine, serine and ferric nitrate
  • Contains four fold concentration of amino acids and vitamins
Dulbecco’s Modified Eagle Medium is one of the most widely used modification of Eagle’s medium. DMEM is a modification of Basal Medium Eagle (BME) that contains four fold concentration of amino acids and vitamins. Additionally, the formulation also includes glycine, serine and ferric nitrate. The original formulation contains 1000 mgs/L of Glucose and was originally used to culture embryonic mouse cells. DMEM high glucose is a further modification of original DMEM and contains 4500 mgs/L of glucose. The additional glucose has proved to be useful in cultivating various other cell lines including primary cultures of mouse and chicken cells as well as various normal and transformed cell lines.