PYREX® Reusable Volumetric Pipettes, Corning
Supplier: Corning
7102-1X 7102-1 7102-2 7102-3 7102-10 7102-20 7102-100 7102-5 7102-25 7102-50
53043-012CS 459.35 CAD
53043-012 53043-034 53043-056 53043-078 53043-170 53043-260 53043-307 53043-114 53043-271 53043-282
PYREX® Reusable Volumetric Pipettes, Corning
Pipettes Graduated Pipettes
These reusable pipettes are designed primarily for schools and laboratories where the Class A tolerance is not required.
  • Larger tip openings for faster delivery
  • Color-coded
  • Colored graduations
  • Accuracy tolerances are twice those specified for Class A volumetric ware
They are constructed to the same wall thickness and length as standard Class A volumetric pipettes, but have larger tip openings for faster delivery. Primarily due to the faster delivery, accuracy tolerances are twice those specified for Class A volumetric ware. Reference: ASTM E-969.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey