PYREX® EZ Access™ Extra-Wide Neck Volumetric Flask, Class A, Corning
Supplier: Corning
5655-50 5655-100 5655-200 5655-250 5655-500 5655-1L
66025-660CS 137.63 CAD
66025-660 66025-662 66025-664 66025-666 66025-668 66025-670
PYREX® EZ Access™ Extra-Wide Neck Volumetric Flask, Class A, Corning
Flasks Volumetric Flasks
These PYREX® EZ Access™ heavy walled volumetric flasks are designed to meet Class A, ASTM tolerances for volumetrics.
  • Calibrated 'to contain' (TC)
  • GL 32 high temperature screw cap fits all flasks
  • Screw cap with PTFE liner provides a secure closure for leak proof storage of solutions
  • Extra large volume size is permanently screened onto the flask body for easy volume identification
The wide neck design allows for easy filling and mixing of various materials into solution and also allows access to larger capacity pipettes.