Green Retractor, OR Grade, Sklar
Supplier: Sklar
57-1485
10197-698EA 290.67 CAD
10197-698
Green Retractor, OR Grade, Sklar
Retractors
Sklar's Green Retractor is a handheld, single-end retractor most commonly used in thyroidectomy and other procedures in the neck
It has a strongly curved, smooth, fenestrated blade. It also has a teardrop fenestrated handle.