Killian Nasal Speculum, OR Grade, Sklar
Supplier: Sklar
70-1020 70-1030 70-1035
10198-506EA 726.39 CAD
10198-506 10198-508 10198-510
Killian Nasal Speculum, OR Grade, Sklar
Specula
Sklar's Killian Nasal Speculum is used for examination of the nasal cavity
This product has a set screw locking mechanism so the speculum may be locked into an open position, therefore freeing the hands of the user. The device has a single spring plier handle, and is available with a small, medium, or large speculum.