Otoscope, OR Grade, Sklar
Supplier: Sklar
67-6250
10198-452EA 478.58 CAD
10198-452
Otoscope, OR Grade, Sklar
Opthalmoscopes
The Sklar Otoscope employs a Rheostat for varied light intensity.
  • Rheostat provided for varied light intensity
  • Three specula
  • Chrome plated
  • Reusable
  • Latex-free
This Otoscope has three specula (sizes 2.5 mm, 3.5 mm, 4.5 mm).
Ordering information: Batteries required, need to be ordered separatly.