Sulfuric acid 19.2 N volumetric solution

Supplier: Hach
Danger

203832 203849
CA2038-32EA 81.69 CAD
CA2038-32 CA11016-366
Sulfuric acid 19.2 N volumetric solution
Sulfuric acid

For determination of volatile acids by the esterification method.

Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR