KineticButler Shoe Cover Dispenser, Protexer
Supplier: Protexer
KBLG KBSM
10842-396EA 3638.93 CAD
10842-396 10842-398
KineticButler Shoe Cover Dispenser, Protexer
PPE Dispensers
The KineticButler Shoe Cover Dispenser is designed for low, moderate, or high volume areas where speed, safety, and compliance are critical. The simple design allows booties to be applied hands-free and enables workers to enter facilities without dropping tools or touching dirty shoes. Dispensers are available in two sizes. The large unit has a handle for safety and stability while applying shoe covers, while the small unit is more easily transported. Both units require no electricity for operation.
  • Designed for low, moderate, or high volume areas where speed, safety, and compliance
More Product Information
Order Now
SPECIFICATIONS
PPE type Shoe covers
Environmentally Preferable
Dispenser type Floor-standing