Glass Vials, C&G Containers
Supplier: C&G Containers
LU23A000A01FZS LFV0403000 LV73A000A01BZS LV74H100B21BZS LAV040700005
71001-232CS 356.92 CAD
71001-232 71001-234 71001-238 71001-240 71001-242 71001-244 71001-246 71001-248 71001-250 71001-252 71001-254 71001-256 71001-258 71001-260 71001-262 71001-264 71001-266 71001-268 71001-270 71001-276 71001-284 71001-288 71001-290 71001-292 71001-294 71001-296 71001-298 71001-300 71001-302 71001-304 71001-306 71001-310 71001-312 71001-314 71001-316 71001-318 71001-320 71001-322 71001-324 71001-326 71001-328 71001-330 71001-332 89137-128 89137-130 89137-132 89137-134 89370-068 30617-868 10146-996 10146-998 10496-920 10496-922 10496-926 10496-928 75840-124
Glass Vials, C&G Containers
Vials
These glass vials from C&G Containers are designed primarily for the collection and testing of solid samples for Volatile Organic Analysis and Total Petroleum Hydrocarbons (TPH), and for the collection and testing of aqueous samples for Total Organic Carbon (TOC).
  • All glass vials are borosilicate Type 1 glass and use a 24-414 closure
Pre-cleaned and Certified products contain a Certificate of Analysis and Cleaning Protocol Form in the product case. Wash protocols for these vials include; A1 (Nitric acid wash) certified, B1 process certified, L3 (no wash, vial with cap attached) not certified, and D1 protocol, a depyrognation-like process for low level Total Organic Carbon (TOC), certified. Some vials are tare weighed. Some vials have dust covers.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Caps can be either one-piece Open Top polypropylene with PTFE septa ultrasonically bonded (not glued), two-piece Open Top polypropylene with pressure fitted PTFE septa, solid top polypropylene cap with PTFE septa, or solid top polypropylene cap with PTFE liner. The PTFE septa is a 3.1 mm fluoropolymer resin/silicone septa laminate (overall thickness 0.125", ±0.006", silicone thickness 0.115", PTFE 0.010"). There is also a thin bonded PTFE Septa cap, overall thickness 0.060 ±0.005" (silicone: 0.056", PTFE: 0.004"). The PTFE liner is a 0.10" thick fluoropolymer disc.
Secondary uses include containment of extracts. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity "contain" volumes may vary.