Finochietto Rib Spreader, OR Grade, Sklar
Supplier: Sklar
55-7680 55-7685
10197-642EA 2506.49 CAD
10197-642 10197-644
Finochietto Rib Spreader, OR Grade, Sklar
Retractors
Sklars® Finochietto rib spreader with two bladed arms is used in thoracic procedures to allow clear access to the surgical field.
  • Available in large and medium sizes
Like the Harken rib spreaders, these also has fenestrated blades, distinguishing it from theTuffier.
Order Now
SPECIFICATIONS
Disposable / reusable Reusable
Environmentally Preferable
Latex/Latex-free Latex-free