Farabeuf Retractor, OR Grade, Sklar
Supplier: Sklar
60-1460 60-1461
10197-268PK 236.83 CAD
10197-268 10197-270
Farabeuf Retractor, OR Grade, Sklar
Retractors
Sklar Farabeuf Double-Ended Retractor is a versatile handheld retractor that is used in multiple procedures
It may be used in dentistry, wrist and hand procedures, or hernia repair, among other applications. The instrument is a solid piece of stainless steel with one angled blade at each end. Each blade is smooth, blunt, and also has a slight upward lip at the end.