PYREX® Ignition Tube, Corning
Supplier: Corning
9860-14 9860-16 9860-25
89091-156CS 1612.64 CAD
89091-156 89091-184 60835-247
PYREX® Ignition Tube, Corning
Tubes Test Tubes
These PYREX® rimless ignition tubes are made from No. 7740 glass.
  • Very good chemical resistance
  • Good temperature resistance
These ignition tubes are satisfactory for most applications encountered in school work where temperatures above 600 °C are not required. These rimless tubes perform well when continually used at temperatures up to 500 °C, or for short periods to 600 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey