ITSE Animal Free®, InVitria
Supplier: Invitria
777ITS032-10ML 777ITS032100ML 777ITS03210X10
10847-714EA 278.73 CAD
10847-714 10847-794 75784-766
ITSE Animal Free®, InVitria
Cell Culture Supplements and Additives
InVitria’s ITS, ITSE, and ITSE+A Animal-Free® are recombinant, blood-free cell culture media supplements used to optimize cell growth.
For optimal proliferation and performance, cells cultured must be supplied with a critical proteins and components:
Insulin: promotes uptake of glucose and amino acids, facilitating cellular signaling
Transferrin: delivers intracellular iron via the transferrin receptor, drives cellular respiration, chelates free iron, and reduces generation of cytotoxic free radicals and peroxide
Selenium: required for the activity of antioxidant enzymes such as glutathione peroxidase and thioredoxin reductase
Ethanolamine: phospholipid precursor that improves the performance of cells in serum-free media and is required for the growth of certain cell types
Albumin: binds, transports, and delivers ions, fatty acids, vitamins, hormones, small-molecule drugs, and toxins; regulates serum pH and osmotic pressure; and provides antioxidant activity