SMART Digest™ Protein Digests Kits

Supplier: Thermo Scientific

60109-101 60109-102 60109-103 60109-101-B 60109-101-MB 60109-102-B 60109-103-B 60109-103-MB 60109-104-B 60109-104-MB 60109-105-B 60109-106-B 60109-106-MB 60109-107 60109-107-B 60109-108 60109-108-B 60109-109 60109-109-B 60109-109-MB 60109-104 60109-105 60109-106 60109-105-MB 60109-107-MB 60109-108-MB 60113-101 60109-102-MB 60109-101-LPH
10790-384EA 1655.09 CAD
10790-384 10790-386 10790-388 10017-750 75838-870 75838-872 75838-874 75838-876 75838-878 75838-880 75838-882 75838-884 75838-888 75838-890 75838-892 75838-894 75838-896 75838-898 75838-900 75838-902 75839-400 75839-402 75839-404 75839-426 75839-428 75839-430 75839-564 75839-574 76005-236 76312-780
SMART Digest™ Protein Digests Kits
Protein Extraction Protein Extraction Reagents

Obtain high quality analytical results from protein digests by using Thermo Scientific™ SMART Digest™ Kits.


  • Increased speed
  • Three-step sample preparation
  • High reproducibility
  • Higher sequence coverage
  • Compatible with automation


A significant advance in sample preparation for biopharmaceutical and protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation.


Significantly increased speed of sample processing with 3-step sample preparation. Significantly reduces sample preparation time, typically takes less than 60 minutes (dependant on sample complexity) to achieve full digestion. Higher sequence coverage is due to the removal of the reduction and alkylation steps and absence of trypsin. This reduces the number of chemicals and presence of trypsin in the final sample, as well as fewer post translational modifications, resulting in lower suppression of signal during LC-MS analysis.


For use with: Peptide quantitation, peptide mapping.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR