CTC LEAP CombiPAL, HDHT Headspace Syringe, Hamilton
Supplier: Hamilton
209681 209682 209683 209684 209685 209686
10836-936EA 408 CAD
10836-936 10836-938 10836-940 10836-942 10837-054 75778-158
CTC LEAP CombiPAL, HDHT Headspace Syringe, Hamilton
Syringes Syringes for Chromatography
Hamilton introduces a new headspace syringe featuring a unique Glue-Free (GF) needle attachment in combination with the well-known High Dynamic (HD) plunger designed specifically for PAL COMBI-xt Headspace Autosamplers.
  • Glue-free needle
  • High dynamic (HD) plunger
  • Chemically inert
Glue-Free needle attachment is chemically inert, eliminating detached needles due to contact with organic, and chlorinated solvents. Glue-Free needle attachment minimizes ghost peaks. Temperature stability up to 200°C means a wider range of sample components can be analyzed.More Product Information