Borosilicate Glass Beads, Chemglass
Supplier: Chemglass
CG-1101-05 CG-1101-01 CG-1101-02 CG-1101-03 CG-1101-04 CG-1101-06
80066-212EA 448.16 CAD
80066-212 80066-204 80066-206 80066-208 80066-210 80066-214
Borosilicate Glass Beads, Chemglass
Beads Glass Beads
Solid borosilicate spheres.
  • Beads have excellent chemical resistance
  • Can be used as distillation column packing
  • Can be used as boiling stones
  • Can be used to provide support for chromatography columns used in gel filtrations
Packaging: Packed in boxes containing one pound.