P-Vac Portable Vacuum System, Argos Technologies
Supplier: Argos Technologies
4397-07
10847-282EA 1287.18 CAD
10847-282
P-Vac Portable Vacuum System, Argos Technologies
Pumps Vacuum Aspiration Systems
The P-Vac has been designed as a compact, flexible and portable liquid waste disposal system
The P-Vac can be operated with bottle top aspirators, or any glass or plastic bottles up to 2 liters. The unit is supplied with a quiet, adjustable diaphragm pump, which can empty a 2 liter container in just 40 seconds with an aspirating pipette. The hydrophobic filter protects the unit from over-filling.
The unit’s small footprint makes it ideal for the laboratory bench or laminar hood. It has been designed to accommodate a variety of collection vessels including bottle top aspirating filter units, plastic vacuum bottles, glass Erlenmeyer flasks, or any device that accepts vacuum. The rechargeable lithium ion battery allows for use where an inline vacuum source is not available, outside of the laboratory setting where a vacuum is required, or when the sample being aspirated is too dangerous for the house vacuum.
Ordering information: The P-Vac system is supplied with a .45um hydrophobic vacuum filter, silicone connection tubing, HandE-Vac hand operator with single channel pipette tip adapter and a universal main adapter.