Tetrakis(triphenylphosphine)palladium(0) ≥ 99.5% (≥ 9% Pd, Assay)
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
Packaged under argon.
04833-1G
04833-25G
04833-5G
CH04833-1GEA
59.03
CAD
CH04833-1G
CH04833-25G
CH04833-5G
Tetrakis(triphenylphosphine)palladium(0) ≥ 99.5% (≥ 9% Pd, Assay)
Tetrakis(triphenylphosphine)palladium (0)
Pd(PPh3)4 is widely used as a catalyst for palladium-catalyzed coupling reactions.
Prominent applications include the Heck reaction, Suzuki coupling, Stille coupling, Sonogashira coupling, and Negishi coupling.
Store at 0-8 °C under Argon.
Formula:
C₇₂H₆₀P₄Pd MW: 1155.58 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 |
Specification Test Results
Appearance | Yellow Solid |
Infrared Spectrum | Conforms to Structure |
Assay | ≥99.5% (≥9% Pd) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...