PEEK Luer Adapters, 1-Way, Cadence Science®
Supplier: Cadence Science
3600 3601 3602 3605 3606 3608 3609
76268-966EA 140.21 CAD
76268-966 76268-968 76268-970 76268-974 76268-976 76268-978 76268-980
PEEK Luer Adapters, 1-Way, Cadence Science®
Fittings and Connectors
Made of inert PEEK and designed for quick, leak-proof connections.
  • Quick leak-proof connections
  • Economical
These adapters are economical and provide for flexibility of a variety of threaded connections.