Double Filtered Balance Enclosures, Labconco®
Supplier: Labconco
3971280 3971281 3971282 3971380 3971381 3971482 3971485 3971486 3971487 3971490 3971491 3971492 3971585 3971586 3971687 3971690 3971691 3971692 3971591 3971592 3971685 3971382 3971480 3971481 3971587 3971590 3971686
10830-828EA 21464.3 CAD
10830-828 10830-830 10830-832 10830-834 10830-836 10830-838 10830-840 10830-844 10830-846 10830-848 10830-850 10830-852 10830-854 10830-856 10830-858 10830-860 10830-862 10830-866 10830-868 10830-870 10830-872 10830-880 10830-882 10830-884 10830-886 10830-888 10830-890
Double Filtered Balance Enclosures, Labconco®
Fume Hoods and Enclosures Balances Enclosures
The patented RXPert Double Filtered Balance System is a Class I Enclosure that meets the USP <800> requirements for non-sterile compounding with NO DUCTWORK! It provides user protection by keeping powders and particulates contained during hazardous drug manipulation
The RXPert features in intrinsically safe negative pressure design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. This balance enclosure includes 2 HEPA filters to provide redundant HEPA filtration in series. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement.
Each of the HEPA filters 100% independently scanned for easy leak detection without removing impellers or filters. It includes a built-in blower with dynamically-balanced, vibration-isolated motorized impeller and speed control. The blower has a 40% reserve so that the speed may be adjusted as needed by a certified technician as the filter loads.
Optional filtration kits and thimble ducting kits are available to convert existing XPert Balance Filtered Systems and XPert Bulk Powder Filtered Systems. RXPert Single Filtered models with factory-installed canopy and damper for ducting outside are also available.
It features durable glass and steel construction, ergonomic air foil with aerodynamic Clean-Sweep airflow openings, and deep interior of 23.4" or 30.0" depending on the model.
Certifications: ETL listed and CE marked. All models conform to the following: SEFA 1-2010; Modified ASHRAE 110-1995; ANSI Z9.5-2011; UL 61010-1; CAN/CSA C22.2 No. 1010.1.
Ordering information: All models include a built-in analog airflow monitor. Widths of 2', 3' and 4' with standard height and 4’, 5’ and 6’ with tall height or tall height and extended depth are available. Some models include a stainless steel or epoxy work surface. A work surface is required if one is not included. All models require a supporting base (not included).