ProKlenz® ONE Alkaline Detergent, STERIS®
Supplier: Steris
142105EC 142101EC 1421D3EC
CA11007-736EA 405.17 CAD
CA11007-736 CA11007-738 CA11007-740
ProKlenz® ONE Alkaline Detergent, STERIS®
Cleaning Agents Detergents and Surface Cleansers
ProKlenz® ONE High performance alkaline cleaner is ideal for use in clean-in-place (CIP), clean-out-of-place (COP) and manual applications.
  • NaOH-based detergent, environmentally friendly
  • Delivers effective microbial control in critical manufacturing processes with its disinfectant and virucidal properties
  • Manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • An extensive support package is available to meet validation objectives
It is a proprietary blend of a concentrated alkalinity source, advanced surfactant system and other performance-enhancing ingredients provide multiple cleaning mechanisms
It is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, nutraceutical, and other industries requiring an effective validatable cleaner.
This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants.
Certifications: Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.