Anti-CD56 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5.1H11]

Supplier: Biolegend
362507 362508
CA10760-860EA 186.71 CAD
CA10760-860 CA10760-862
Anti-CD56 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5.1H11]
Antibodies
PE anti-human CD56 (NCAM) [5.1H11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.4 μg per million cells in 100 μl volume. For flow cytometric staining using test sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: 5.1H11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR