Anti-CD5 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 53-7.3]

Supplier: Biolegend
100617 100618
CA10756-680EA 290.44 CAD
CA10756-680 CA10756-682
Anti-CD5 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 53-7.3]
Antibodies
Brilliant Violet 421™ anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the ug size, the suggested use of this reagent is 0.03 μg per million cells in 100 μl volume. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Brilliant Violet 421™ excites at 405 nm and emits at 421 nm. The standard bandpass filter 450/50 nm is recommended for detection. Brilliant Violet 421™ is a trademark of Sirigen Group Ltd.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR