Anti-GFAP Mouse Monoclonal Antibody [clone: SMI 26]

Supplier: Biolegend
837601 837602
CA10817-804EA 494.7 CAD
CA10817-804 CA10817-806
Anti-GFAP Mouse Monoclonal Antibody [clone: SMI 26]
Antibodies
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: SMI 26
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b/IgG1
Reactivity: Human, Sheep, Cow, Dog, Pig, Rat, Guinea pig, Rat, Mouse, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR