Anti-PIP Mouse Monoclonal Antibody [clone: D6]

Supplier: Biolegend
915101 915102 915103
CA10817-514EA 1005.35 CAD
CA10817-514 CA10817-516 CA10817-518
Anti-PIP Mouse Monoclonal Antibody [clone: D6]
Antibodies
Type: Primary
Antigen: PIP
Clonality: Monoclonal
Clone: D6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR