Anti-APP Mouse Monoclonal Antibody [clone: 4G8]

Supplier: Biolegend
800709 800710 800711
CA10816-388EA 1681.97 CAD
CA10816-388 CA10816-390 CA10816-392
Anti-APP Mouse Monoclonal Antibody [clone: 4G8]
Antibodies
Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 4G8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR