Anti-APP Mouse Monoclonal Antibody [clone: 4G8]

Supplier: Biolegend
800704 800705 800706
CA10816-380EA 1043.65 CAD
CA10816-380 CA10816-382 CA10816-384
Anti-APP Mouse Monoclonal Antibody [clone: 4G8]
Antibodies
Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 4G8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR