Anti-TRAF6 Mouse Monoclonal Antibody [clone: T2-1SC]

Supplier: Biolegend
654501 654502
CA10763-504EA 253.73 CAD
CA10763-504 CA10763-506
Anti-TRAF6 Mouse Monoclonal Antibody [clone: T2-1SC]
Antibodies
Purified anti-TRAF6 [T2-1SC]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TRAF6
Clonality: Monoclonal
Clone: T2-1SC
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR