Anti-TNF-a Rat Monoclonal Antibody [clone: MP6-XT22]

Supplier: Biolegend
506331 506332
CA506331-BLEA 290.44 CAD
CA506331-BL CA506332-BL
Anti-TNF-a Rat Monoclonal Antibody [clone: MP6-XT22]
Antibodies
Ultra-LEAF™ Purified anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA, ICFC, CyTOF®, IF, IHC, WB, Neut; Size: 1 mg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of mouse TNF-α recombinant protein ranging from 500 to 4 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR