Anti-TNF-a Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: MAb11]

Supplier: Biolegend
502935 502936
CA502935-BLEA 386.18 CAD
CA502935-BL CA10761-952
Anti-TNF-a Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: MAb11]
Antibodies
Brilliant Violet 605™ anti-human TNF-α [MAb11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine), Pigtailed Macaque, Sooty Mangabey; Apps: ICFC, FC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MAb11
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Swine (Pig;Porcine);Pigtailed Macaque;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR