Anti-TCR-b chain Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: H57-597]

Supplier: Biolegend
109229 109230
CA109229-BLEA 280.86 CAD
CA109229-BL CA10756-340
Anti-TCR-b chain Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: H57-597]
Antibodies
Brilliant Violet 421™ anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performanceMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol TCR-b chain TCR-b chain
Environmentally Preferable
Antigen synonyms TCRB|β-TCR|TCR-β chain|TCR-β TCRB|β-TCR|TCR-β chain|TCR-β
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation Brilliant Violet® 421 Brilliant Violet® 421
Clone H57-597 H57-597
Reactivity Mouse Mouse
Host Armenian hamster Armenian hamster
Gene ID 21577 21577
Isotype IgG IgG
Immunogen Affinity purified TCR from mouse DO-11.10 cells Affinity purified TCR from mouse DO-11.10 cells
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA). Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody. The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze. The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR